Skip to Content
Merck
All Photos(1)

Key Documents

428736

Supelco

Amberlite MB20 Ion Exchange Resin

mixed bed, >50 mesh

Synonym(s):

AmberLite MB20 H/OH Mixed Bed Ion Exchange Resin hydrogen and hydroxide form

Sign Into View Organizational & Contract Pricing

Select a Size

1 KG
946,00 kr.

946,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
1 KG
946,00 kr.

About This Item

MDL number:
UNSPSC Code:
12000000
NACRES:
SB.52

946,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Product Name

AmberLite MB20 Mixed Ion Exchange Resin,

form

beads

Quality Level

composition

HCR-S[H+] and SBR[OH-]: in accordance

parameter

60 °C max. temp.

moisture

46-51% (cation exchanger)
60-72% (anion exchanger)

technique(s)

LPLC: suitable

matrix

styrene-divinylbenzene (gel)

particle size

20-50 mesh
560-660 μm (anion form)
710-810 μm (cation form)

operating pH

0-14

capacity

1.0 meq/mL by wetted bed volume(anion)
1.9 meq/mL by wetted bed volume

separation technique

mixed mode

Looking for similar products? Visit Product Comparison Guide

General description

AmberLite MB20 is a premium, premixed, mixed bed product designed for polishing demineralized water to conductivities <0.1 µS/cm.

Application

AmberLite MB20 hydrogen and hydroxide form was used in assessing inorganic nitrogen in litter layer and soil using ion-exchange resin bags on Alpkem autoanalyzer [1] and by ion chromatography. [2] Mixed bed ion exchange resin specially processed resin for condensate polishing; best for in-process and column demineralization.
Mixed bed ion exchange resin specially processed resin for condensate polishing; best for in-process and column demineralization.

Features and Benefits

Uniform size beads offer higher surface area for increased capacity and faster kinetics. Nuclear grade, gel form, styrene-divinylbenzene copolymer matrix

Other Notes

This ion-exchange resin has not been specially processed nor cleaned. We suggest that it be treated to a suitable preliminary elution and wash.

Legal Information

Amberlite is a trademark of DuPont de Nemours, Inc.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Functional diversity indices reveal the impacts of land use intensification on plant community assembly.
Pakeman, Robin J.
Journal of Ecology, 99, 1143-1151 (2011)
Contrasting effects of substrate and fertilizer nitrogen on the early stages of litter decomposition.
Hobbie, Sarah E.
Ecosystems, 8, 644-656 (2005)

Questions

  1. Questa resina permette di correggere pH estremamente acidi o basici e portarli verso il pH 7?

    1 answer
    1. Depending on whether cations or anions are put through the resin, this material will release H+ ions (if cations are in solution) or OH- ions (if anions are in solution). That said, it can raise or lower the pH, depending on the solution being used, but it is not inherently used to regulate pH; it depends upon the solution properties. The resin is typically used for demineralization and polishing applications rather than pH control.

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service