Skip to Content
Merck
All Photos(1)

Documents

40982

Supelco

Maize GMO Standard ERM-BF423

Set for 0%, 0.1%, 1%, 10% MIR604

Synonym(s):

GVO Standard ERM-BF423, Maize MIR604

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107

product line

BioChemika

application(s)

cleaning products
cosmetics
food and beverages
microbiology
personal care
pharmaceutical (small molecule)

format

matrix material

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

12 - Non Combustible Liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service