Skip to Content
Merck
All Photos(1)

Key Documents

39421

Sigma-Aldrich

4-(Dimethylamino)cinnamaldehyde

≥98.0% (HPLC)

Synonym(s):

DMAC, DMACA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2NC6H4CH=CHCHO
CAS Number:
Molecular Weight:
175.23
Beilstein:
972369
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:

Assay

≥98.0% (HPLC)

mp

138-140 °C (lit.)
138-141 °C

solubility

chloroform/ethanol (1:1): 50 mg/mL, clear to very faintly turbid, yellow to deep red

SMILES string

CN(C)c1ccc(\C=C\C=O)cc1

InChI

1S/C11H13NO/c1-12(2)11-7-5-10(6-8-11)4-3-9-13/h3-9H,1-2H3/b4-3+

InChI key

RUKJCCIJLIMGEP-ONEGZZNKSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

Reagent for the determination of aromatic amines; e.g. PABA

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A simple assay for measurement of urinary p-aminobenzoic acid in the oral pancreatic function test.
C Yamato et al.
Analytical biochemistry, 98(1), 13-17 (1979-09-15)
M. Qureshi et al.
Analytica Chimica Acta, 86, 309-309 (1976)
Mark J Payne et al.
Journal of AOAC International, 93(1), 89-96 (2010-03-26)
A simple, specific, high-throughput colorimetric method based on the reaction of 4-dimethylaminocinnamaldehyde (DMAC) with flavan-3-ols was developed to determine total procyanidins in selected cacao-based products. Extracts of defatted samples were dispensed into a 96-well plate and reacted with DMAC. The
Sharon Abrahams et al.
Plant physiology, 130(2), 561-576 (2002-10-12)
Proanthocyanidin (PA), or condensed tannin, is a polymeric flavanol that accumulates in a number of tissues in a wide variety of plants. In Arabidopsis, we found that PA precursors (detected histochemically using OsO(4)) accumulate in the endothelial cell layer of
C L Stone et al.
The Journal of biological chemistry, 268(2), 892-899 (1993-01-15)
Human liver alcohol dehydrogenase isoenzymes beta 1 beta 1 and beta 2 beta 2, in which position 47 in the coenzyme binding domain is an arginine or histidine, respectively, differ remarkably in steady-state kinetics. To understand which catalytic steps affect

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service