Skip to Content
Merck
All Photos(1)

Key Documents

38030

Sigma-Aldrich

3,5-Diiodosalicylic acid

purum, ≥98.0% (T)

Synonym(s):

2-Hydroxy-3,5-diiodobenzoic acid, 3,5-Diiodo-2-hydroxybenzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
I2C6H2(OH)CO2H
CAS Number:
Molecular Weight:
389.91
Beilstein:
2615358
EC Number:
MDL number:
UNSPSC Code:
12352100
NACRES:
NA.22

Pricing and availability is not currently available.

grade

purum

Assay

≥98.0% (T)

ign. residue

≤0.1%

mp

220-230 °C (dec.) (lit.)

SMILES string

OC(=O)c1cc(I)cc(I)c1O

InChI

1S/C7H4I2O3/c8-3-1-4(7(11)12)6(10)5(9)2-3/h1-2,10H,(H,11,12)

InChI key

DHZVWQPHNWDCFS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

A soluble surfactant

replaced by

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Phospholipase A2 activity on mixed lipid monolayers: inhibition and activation of phospholipases A2 from porcine pancreas and Crotalus adamanteus by anionic and neutral amphiphiles.
H S Hendrickson et al.
Archives of biochemistry and biophysics, 212(2), 508-514 (1981-12-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service