Skip to Content
Merck
All Photos(1)

Documents

37035

Supelco

Universal Indicator paper pH 1-11

Triple color chart

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41113319

Quality Level

feature

non-bleeding: no
refill: no

suitability

passes test for shade of color acc. to scale

application(s)

food and beverages
general analytical

detection method

colorimetric

Looking for similar products? Visit Product Comparison Guide

Packaging

Original box with triple color chart
roll with approx. 5 m

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A precise method for the analysis of δ18O of dissolved inorganic phosphate in seawater.
McLaughlin, Karen, et al.
Limnology and Oceanography: Methods, 2, 202-212 (2004)
Denise Walker
Acids and Alkalis, 13-13 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service