Skip to Content
Merck
All Photos(1)

Documents

36848

Supelco

Epoxiconazole

PESTANAL®, analytical standard, Racemate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H13ClFN3O
CAS Number:
Molecular Weight:
329.76
Beilstein:
9509539
EC Number:
UNSPSC Code:
41116107
PubChem Substance ID:

grade

analytical standard

product line

PESTANAL®

quality

Racemate

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
cleaning products
cosmetics
environmental
food and beverages
personal care

format

neat

SMILES string

Fc1ccc(cc1)[C@@]3(Cn2cncn2)O[C@@H]3c4ccccc4Cl

InChI

1S/C17H13ClFN3O/c18-15-4-2-1-3-14(15)16-17(23-16,9-22-11-20-10-21-22)12-5-7-13(19)8-6-12/h1-8,10-11,16H,9H2/t16-,17+/m0/s1

InChI key

ZMYFCFLJBGAQRS-DLBZAZTESA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Carc. 2 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C Deweer et al.
Communications in agricultural and applied biological sciences, 77(3), 109-115 (2012-01-01)
DMIs have been for many years the most used antifungal compounds against Septoria tritici blotch caused by Mycosphaerella graminicola, the most important foliar disease on winter wheat worldwide. Their use leads to a development and widespread of isolates with reduced
Ivan Rybár et al.
Journal of separation science, 30(18), 3164-3173 (2007-11-21)
An analytical method combining off-line flow-through extraction of a soil micro-sample (mass around 100 mg, packed into a short HPLC glass column) and direct on-column large-volume injection (LVI up to 1.00 mL) of a methanol-water soil extract onto a conventional
Camilla Taxvig et al.
Toxicological sciences : an official journal of the Society of Toxicology, 100(2), 464-473 (2007-09-06)
The triazole fungicides tebuconazole and epoxiconazole were investigated for reproductive toxic effects after exposure during gestation and lactation. Rats were dosed with epoxiconazole (15 or 50 mg/kg bw/day) or tebuconazole (50 or 100 mg/kg bw/day) during pregnancy from gestational day
F Henriet et al.
Communications in agricultural and applied biological sciences, 71(2 Pt A), 193-195 (2007-03-30)
Three trials were carried out during springs 2003 and 2004 to compare the distribution of fungicides on the different leaf layers of wheat plants. Mixtures of 1 L/ha of Amistar (SC, 250 g/L of azoxystrobin) and 1 L/ha of Opus
Vera L S S de Castro et al.
Birth defects research. Part B, Developmental and reproductive toxicology, 95(2), 123-129 (2011-12-06)
Although some studies have pointed out to embryo/fetal toxicity, knowledge about the potential toxicity of the fungicide epoxiconazole is still limited. Once the results of these previous studies have raised some concern, this study studied the effects of epoxiconazole maternal

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service