Skip to Content
Merck
All Photos(1)

Documents

34559

Supelco

Imazalil-(allyl-d5)

PESTANAL®, analytical standard

Synonym(s):

1-[2-(Allyl-d5-oxy)-2-(2,4-dichlorophenyl)ethyl]imidazole, Imazalil-d5

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14D5H9Cl2N2O
CAS Number:
Molecular Weight:
302.21
MDL number:
UNSPSC Code:
77101502
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

application(s)

agriculture
environmental

format

neat

SMILES string

ClC1=CC(Cl)=CC=C1C(OC([2H])([2H])C([2H])=C([2H])[2H])CN2C=NC=C2

InChI

1S/C14H14Cl2N2O/c1-2-7-19-14(9-18-6-5-17-10-18)12-4-3-11(15)8-13(12)16/h2-6,8,10,14H,1,7,9H2/i1D2,2D,7D2

InChI key

PZBPKYOVPCNPJY-MACMZUOASA-N

Application


  • Monitoring pesticide residues in olive oil: A study detailed the use of LC-MS/MS for monitoring seven pesticide residues, including Imazalil, in extra virgin olive oil, assessing consumer risk exposure and emphasizing the method′s accuracy and sensitivity for food safety (Schiano et al., 2024).

  • Antifungal properties of modified chitosan: This study synthesized maleopimaric anhydride-modified chitosan and investigated its antifungal properties, including against fungi susceptible to Imazalil, suggesting potential applications in biodegradable antifungal materials (Li et al., 2024).

  • Evaluation of fungicide mixtures in fish toxicity: An investigation into the toxic mechanisms of Imazalil and azoxystrobin, alone and in combination, on hook snout carp, provides insights into the toxic dynamics and safety thresholds needed for aquatic life conservation in environments exposed to these fungicides (Lv et al., 2024).

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Dam. 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service