Skip to Content
Merck
All Photos(2)

Key Documents

34193

Supelco

Dioctyl phthalate-3,4,5,6-d4

analytical standard

Sign Into View Organizational & Contract Pricing

Select a Size

25 MG
1.560,00 kr.

1.560,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
25 MG
1.560,00 kr.

About This Item

Linear Formula:
C6D4[CO2CH2(CH2)6CH3]2
CAS Number:
Molecular Weight:
394.58
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

1.560,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.486 (lit.)

bp

384 °C (lit.)

mp

−50 °C (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

mass shift

M+4

SMILES string

[2H]c1c([2H])c([2H])c(C(=O)OCCCCCCCC)c(c1[2H])C(=O)OCCCCCCCC

InChI

1S/C24H38O4/c1-3-5-7-9-11-15-19-27-23(25)21-17-13-14-18-22(21)24(26)28-20-16-12-10-8-6-4-2/h13-14,17-18H,3-12,15-16,19-20H2,1-2H3/i13D,14D,17D,18D

InChI key

MQIUGAXCHLFZKX-AMEAAFLOSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

related product

Product No.
Description
Pricing

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

228.2 °F - closed cup

Flash Point(C)

109 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service