Skip to Content
Merck
All Photos(1)

Key Documents

33224

Sigma-Aldrich

Glycerol solution

puriss. p.a., reag. ISO, reag. Ph. Eur., ≥86%

Synonym(s):

1,2,3-Propanetriol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOCH2CH(OH)CH2OH · aq
CAS Number:
Molecular Weight:
92.09 (anhydrous basis)
Beilstein:
635685
EC Number:
MDL number:
UNSPSC Code:
12190000
PubChem Substance ID:

Agency

reag. ISO
reag. Ph. Eur.

grade

puriss. p.a.

Assay

≥86%

impurities

acidity or alkalinity, complies
aldehyde, complies
impurities A and related substances (GC), complies
sugar, complies
≤0.0001% heavy metals (as Pb)
≤0.0005% ammonium (NH4)
≤0.0005% free alkali (as NH3)
≤0.002% free acid (as CH3COOH)
≤0.003% reducing matter
≤0.005% other organic compounds
≤0.05% fatty acid ester

ign. residue

≤0.005% (as SO4)

color

APHA: >10

refractive index

n20/D 1.4520-1.4550

bp

182 °C/20 mmHg (lit.)

density

1.224-1.230 g/mL at 20 °C
1.252 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤1 mg/kg
sulfate (SO42-): ≤5 mg/kg

cation traces

As: ≤0.5 mg/kg
Fe: ≤0.5 mg/kg

suitability

complies for appearance of solution
complies for reaction against H2SO4
passes test for identity

SMILES string

OCC(O)CO

InChI

1S/C3H8O3/c4-1-3(6)2-5/h3-6H,1-2H2

InChI key

PEDCQBHIVMGVHV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

A -D suffix exists for administrative purposes only.
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hovsep Mahdessian et al.
Proceedings of the National Academy of Sciences of the United States of America, 111(24), 8913-8918 (2014-06-14)
Genome-wide association studies have identified a locus on chromosome 19 associated with plasma triglyceride (TG) concentration and nonalcoholic fatty liver disease. However, the identity and functional role of the gene(s) responsible for these associations remain unknown. Of 19 expressed genes
Shelley L Forrest et al.
The Journal of comparative neurology, 522(17), 3900-3927 (2014-07-22)
Artemin is a member of the glial cell line-derived neurotrophic factor (GDNF) family that has been strongly implicated in development and regeneration of autonomic nerves and modulation of nociception. Whereas other members of this family (GDNF and neurturin) primarily target
Tian Qiu et al.
Nature communications, 5, 5119-5119 (2014-11-05)
Biological microorganisms swim with flagella and cilia that execute nonreciprocal motions for low Reynolds number (Re) propulsion in viscous fluids. This symmetry requirement is a consequence of Purcell's scallop theorem, which complicates the actuation scheme needed by microswimmers. However, most
Marc Simard et al.
The Journal of clinical endocrinology and metabolism, 100(1), E129-E139 (2014-10-17)
Corticosteroid-binding globulin (CBG) is encoded by SERPINA6. It is the major plasma binding protein of glucocorticoids and regulates plasma cortisol levels and bioavailability in humans. Several proteases target CBG and disrupt its steroid-binding properties. To date, most genetic deficiencies that
Nazish Ahmed et al.
Tissue engineering. Part A, 20(15-16), 2224-2233 (2014-03-13)
One of the factors preventing clinical application of regenerative medicine to degenerative cartilage diseases is a suitable source of cells. Chondrocytes, the only cell type of cartilage, grown in vitro under culture conditions to expand cell numbers lose their phenotype

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service