Skip to Content
Merck
All Photos(1)

Key Documents

32359

Supelco

Toltrazuril sulfon-(N-methyl-d3)

VETRANAL®, analytical standard

Synonym(s):

1-(Methyl-d3)-3-{3-methyl-4-[4-(trifluoromethylsulfonyl)phenoxy]phenyl}-1,3,5-triazine-2,4,6(1H,3H,5H)-trione, Ponazuril-d3

Sign Into View Organizational & Contract Pricing

Select a Size

10 MG
4.690,00 kr.

4.690,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 MG
4.690,00 kr.

About This Item

Empirical Formula (Hill Notation):
C18D3H11F3N3O6S
CAS Number:
Molecular Weight:
460.40
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24

4.690,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

grade

analytical standard

Quality Level

product line

VETRANAL®

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
veterinary

format

neat

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service