Skip to Content
Merck
All Photos(1)

Key Documents

32356

Supelco

Hydroxyfenbendazole-(methyl-d3) monohydrate

VETRANAL®, analytical standard

Synonym(s):

N-[6-[(4-Hydroxyphenyl)thio]-1H-benzimidazol-2-yl]carbamic acid methyl-d3 ester monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15D3H10N3O3S · H2O
CAS Number:
Molecular Weight:
336.38
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

product line

VETRANAL®

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

forensics and toxicology
veterinary

format

neat

SMILES string

OC(C=C1)=CC=C1SC2=CC=C(NC(NC(OC([2H])([2H])[2H])=O)=N3)C3=C2.O

InChI

1S/C15H13N3O3S.H2O/c1-21-15(20)18-14-16-12-7-6-11(8-13(12)17-14)22-10-4-2-9(19)3-5-10;/h2-8,19H,1H3,(H2,16,17,18,20);1H2/i1D3;

InChI key

JFFKWYDCSIKODM-NIIDSAIPSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Legal Information

VETRANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service