32002
Sodium 5,5-diethylbarbiturate
purum, ≥99.0% (NT)
Synonym(s):
5,5-Diethylbarbituric acid sodium salt, Barbitone, Sodium barbital, Veronal sodium salt
About This Item
Recommended Products
grade
purum
Quality Level
Assay
≥99.0% (NT)
form
powder or crystals
drug control
regulated under CDSA - not available from Sigma-Aldrich Canada; psicótropo (Spain); Decreto Lei 15/93: Tabela IV (Portugal)
storage condition
(Tightly closed. Dry.)
technique(s)
GC/MS: suitable
HPLC: suitable
activity assay: suitable
color
colorless
pH
9.0-10.5
mp
>287.1 °C (> 548.8 °F)
solubility
water: soluble 103.1 g/L at 20 °C (68 °F)
density
1.418 g/cm3 at 20 °C ( 68 °F)
suitability
suitable for microbiology
suitable for molecular biology
application(s)
cell analysis
SMILES string
[Na+].CCC1(CC)C(=O)NC([O-])=NC1=O
InChI
1S/C8H12N2O3.Na/c1-3-8(4-2)5(11)9-7(13)10-6(8)12;/h3-4H2,1-2H3,(H2,9,10,11,12,13);/q;+1/p-1
InChI key
RGHFKWPGWBFQLN-UHFFFAOYSA-M
Looking for similar products? Visit Product Comparison Guide
General description
Application
Biochem/physiol Actions
Features and Benefits
- Suitable for electrophoresis and enzymatic assays
- High purity product for research applications
- Component of Michaeli′s buffer
Warning
Other Notes
comparable product
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT SE 2 - STOT SE 3
Target Organs
Nervous system, Respiratory system
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service