Skip to Content
Merck
All Photos(1)

Key Documents

29937

Supelco

10-Methoxycarbamazepine

analytical standard

Synonym(s):

10-Methoxy-5H-dibenz[b,f]azepine-5-carboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H14N2O2
CAS Number:
Molecular Weight:
266.29
Beilstein:
1540067
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.24
Product 29937 is not currently sold in your country. Contact Technical Service

grade

analytical standard

Assay

≥97.5% (HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

impurities

≤1.0% water

application(s)

forensics and toxicology
pharmaceutical (small molecule)

format

neat

InChI

1S/C16H14N2O2/c1-20-15-10-11-6-2-4-8-13(11)18(16(17)19)14-9-5-3-7-12(14)15/h2-10H,1H3,(H2,17,19)

InChI key

PIZOFBKQWNPKDK-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service