Skip to Content
Merck

29062

Bis(1,5-cyclooctadiene)dirhodium(I) dichloride

Synonym(s):

1,5-Cyclooctadienerhodium(I) chloride dimer, Chloro(1,5-cyclooctadiene)rhodium(I) dimer, Rhodium(I) chloride 1,5-Cyclooctadiene complex dimer

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C16H24Cl2Rh2
CAS Number:
Molecular Weight:
493.08
MDL number:

mp

~255 °C (dec.) (lit.)

SMILES string

Cl[Rh].Cl[Rh].C1CC=CCCC=C1.C2CC=CCCC=C2

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service