Skip to Content
Merck
All Photos(1)

Key Documents

288667

Sigma-Aldrich

Magnesium oxide

heavy

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
MgO
CAS Number:
Molecular Weight:
40.30
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

form

powder

concentration

57-61% Mg (complexometric)

mp

2852 °C (lit.)

SMILES string

O=[Mg]

InChI

1S/Mg.O

InChI key

CPLXHLVBOLITMK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Suitable for TLC

Other Notes

A -D suffix exists for administrative purposes only.
All -D packages are 100% the same product, same quality, same specification as the package sizes previously sold without a -D.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Benjamin Michen et al.
Environmental science & technology, 47(3), 1526-1533 (2013-01-05)
Ceramic filters, working on the depth filtration principle, are known to improve drinking water quality by removing human pathogenic microorganisms from contaminated water. However, these microfilters show no sufficient barrier for viruses having diameters down to 20 nm. Recently, it
Charles T Campbell et al.
Journal of the American Chemical Society, 134(43), 18109-18115 (2012-10-05)
Adsorbed molecules are involved in many reactions on solid surface that are of great technological importance. As such, there has been tremendous effort worldwide to learn how to predict reaction rates and equilibrium constants for reactions involving adsorbed molecules. Theoretical
Ali Jebali et al.
Toxicology in vitro : an international journal published in association with BIBRA, 27(6), 1896-1904 (2013-06-29)
Cutaneous leishmaniasis (CL) is endemic in the tropical and subtropical countries. Antileishmanial drugs that are traditionally used for treatment of CL are mainly toxic, ineffective for some parasite isolates, and mostly expensive. Previous studies showed that some metal and metal
Magdalena Maciejewska et al.
Journal of chromatography. A, 1257, 141-148 (2012-08-22)
Inverse gas chromatography (IGC) was applied at infinite dilution to evaluate the surface properties of calcium and magnesium oxide nanoparticles and the effect of surface grafted unsaturated carboxylic acid on the nanopowder donor-acceptor characteristics. The dispersive components (γ(s)(D)) of the
Sanket A Deshmukh et al.
Physical chemistry chemical physics : PCCP, 14(44), 15593-15605 (2012-10-19)
Atomic scale characterization of the structure and dynamics of confined water molecules located near the metal oxide-aqueous interface is carried out using molecular dynamics simulations. Proximity effects on water molecules (H(2)O) near a magnesium oxide surface (MgO(100)) at room temperature

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service