258091
3,3′-Diethyloxadicarbocyanine iodide
99%
Synonym(s):
3-Ethyl-2-[5-(3-ethyl-2-benzoxazolinylidene)-1,3-pentadienyl]benzoxazolium iodide, DODC Iodide
About This Item
Recommended Products
Assay
99%
form
powder or crystals
mp
232 °C (dec.) (lit.)
λmax
582 nm
fluorescence
λex 582 nm; λem 603 nm
application(s)
diagnostic assay manufacturing
hematology
histology
storage temp.
room temp
SMILES string
[I-].CCN1\C(Oc2ccccc12)=C\C=C\C=C\c3oc4ccccc4[n+]3CC
InChI
1S/C23H23N2O2.HI/c1-3-24-18-12-8-10-14-20(18)26-22(24)16-6-5-7-17-23-25(4-2)19-13-9-11-15-21(19)27-23;/h5-17H,3-4H2,1-2H3;1H/q+1;/p-1
InChI key
CLDZYSUDOQXJOU-UHFFFAOYSA-M
Looking for similar products? Visit Product Comparison Guide
General description
Application
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service