Skip to Content
Merck
All Photos(1)

Documents

25395

Sigma-Aldrich

5-Chloro-2-nitroaniline

technical, ≥98.0% (HPLC)

Synonym(s):

2-Amino-4-chloro-nitrobenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H3(NO2)NH2
CAS Number:
Molecular Weight:
172.57
Beilstein:
2210201
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

technical

Assay

≥98.0% (HPLC)

mp

125-129 °C (dec.) (lit.)

SMILES string

Nc1cc(Cl)ccc1[N+]([O-])=O

InChI

1S/C6H5ClN2O2/c7-4-1-2-6(9(10)11)5(8)3-4/h1-3H,8H2

InChI key

ZCWXYZBQDNFULS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 2 Oral - Aquatic Chronic 2 - STOT RE 2

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Elena Tibaldi et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 34(1), 1122-1135 (2020-01-10)
Osteopontin (OPN) is a phosphoglycoprotein secreted into the extracellular matrix upon liver injury, acting as a cytokine stimulates the deposition of fibrillary collagen in liver fibrogenesis. In livers of mice subjected to bile duct ligation (BDL) and in cultured activated

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service