Skip to Content
Merck
All Photos(1)

Documents

21886

Sigma-Aldrich

5(6)-Carboxy-2′,7′-dichlorofluorescein diacetate N-succinimidyl ester

for fluorescence, ≥85% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C29H17Cl2NO11
CAS Number:
Molecular Weight:
626.35
MDL number:
UNSPSC Code:
12352200

grade

for fluorescence

Assay

≥85% (HPLC)

mp

≥220 °C (lit.)

solubility

DMF: soluble
DMSO: soluble
methanol: soluble

fluorescence

λex 470 nm; λem 529 nm in 0.1 M Tris pH 8.0 (esterase)

storage temp.

−20°C

SMILES string

CC(=O)Oc1cc2Oc3cc(OC(C)=O)c(Cl)cc3C4(OC(=O)c5cc(ccc45)C(=O)ON6C(=O)CCC6=O)c2cc1Cl.CC(=O)Oc7cc8Oc9cc(OC(C)=O)c(Cl)cc9C%10(OC(=O)c%11ccc(cc%10%11)C(=O)ON%12C(=O)CCC%12=O)c8cc7Cl

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Other Notes

Amine reactive label for long term tracing of cells. Forms green fluorescent conjugates upon deacetylation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C M Griffith et al.
Development (Cambridge, England), 116(4), 1087-1099 (1992-12-01)
During the fusion of rodent embryo palatal shelves, the cells of the outer epithelial layer slough off, allowing the cells of the medial edge basal layer to form a midline seam that undergoes epithelial-mesenchymal transformation, as judged by electron microscopy

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service