Skip to Content
Merck
All Photos(1)

Key Documents

21370

Sigma-Aldrich

(±)-Camphor-10-sulfonic acid

purum, ≥98.0% (T)

Synonym(s):

(±)-β-Camphorsulfonic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16O4S
CAS Number:
Molecular Weight:
232.30
Beilstein:
3205973
EC Number:
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:

grade

purum

Assay

≥98.0% (T)

form

solid

impurities

≤1% water

mp

199-201 °C (dec.)

SMILES string

CC1(C)[C@@H]2CC[C@@]1(CS(O)(=O)=O)C(=O)C2

InChI

1S/C10H16O4S/c1-9(2)7-3-4-10(9,8(11)5-7)6-15(12,13)14/h7H,3-6H2,1-2H3,(H,12,13,14)/t7-,10-/m1/s1

InChI key

MIOPJNTWMNEORI-GMSGAONNSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Determination of 10-camphorsulphonates in pharmaceutical formulations by high-performance liquid chromatography.
C Pierron et al.
Journal of chromatography, 511, 367-372 (1990-07-06)
[D-camphor-beta-sulfonic acid as an ion-pair reagent for the determination of biogenic amines and their metabolites in the rat brain by reverse phase high performance liquid chromatography].
X R Xu et al.
Zhongguo yao li xue bao = Acta pharmacologica Sinica, 8(2), 113-117 (1987-03-01)
Chen Liu et al.
Organic letters, 13(10), 2638-2641 (2011-04-23)
A novel ion pair catalyst containing a chiral counteranion can be readily derived by simply mixing cinchona alkaloid-derived diamine with chiral camphorsulfonic acid (CSA). A mixture of 9-amino(9-deoxy)epi-quinine 8 and (-)-CSA was found to be the best catalyst with matching
Kaoru Nobuoka et al.
Physical chemistry chemical physics : PCCP, 9(44), 5891-5896 (2007-11-09)
We directly observe the interaction between 1-butyl-3-methylimidazolium (bmim) or 1-butyl-2,3-dimethylimidazolium (bm(2)im) and the solute, ethyl acrylate (EA), which is the popular dienophile in the Diels-Alder reaction and an H-bonding acceptor, by using specially designed electrospray mass spectrometry. In imidazolium ionic
Bala Kishan Gorityala et al.
Bioorganic & medicinal chemistry letters, 19(11), 3093-3095 (2009-04-29)
A mild and efficient synthesis of pseudoglycosides has been developed using metal free (S)-camphorsulfonic acid. (S)-CSA acts as an excellent catalyst for conversion of 2,4,6-tri-O-acetyl-D-glucal to 2,3-unsaturated O-glycosides. A wide range of biologically active natural products, alcohols and thiols could

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service