Skip to Content
Merck
All Photos(1)

Key Documents

211001

Sigma-Aldrich

Acid Alizarin Violet N

Dye content 50 %

Synonym(s):

Acid Chrome Violet K, Mordant Violet 5

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H11N2NaO5S
CAS Number:
Molecular Weight:
366.32
Colour Index Number:
15670
EC Number:
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
Pricing and availability is not currently available.

composition

Dye content, 50%

λmax

501 nm

SMILES string

[Na+].Oc1ccc(cc1\N=N\c2c(O)ccc3ccccc23)S([O-])(=O)=O


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service