Skip to Content
Merck
All Photos(1)

Documents

17860

Sigma-Aldrich

2-Bromooctanoic acid

purum, ≥97.0% (GC)

Synonym(s):

2-Bromocaprylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)5CH(Br)COOH
CAS Number:
Molecular Weight:
223.11
Beilstein:
1760958
EC Number:
MDL number:
UNSPSC Code:
12352100

grade

purum

Assay

≥97.0% (GC)

refractive index

n20/D 1.471 (lit.)
n20/D 1.471

bp

140 °C/5 mmHg (lit.)

density

1.278 g/mL at 25 °C (lit.)

SMILES string

CCCCCCC(Br)C(O)=O

InChI

1S/C8H15BrO2/c1-2-3-4-5-6-7(9)8(10)11/h7H,2-6H2,1H3,(H,10,11)

InChI key

GTGTXZRPJHDASG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

2-Bromooctanoic acid is a fatty acyl coenzyme A synthetase (FadD) inhibitor. It also inhibits β-oxidation enzymes in animal cells.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N Mayorek et al.
European journal of biochemistry, 182(2), 395-400 (1989-06-15)
The limiting role of diacylglycerol acyltransferase with respect to triacylglycerol synthesis in cultured rat hepatocytes was evaluated by following the inhibition of the overall synthetic flux by 2-bromooctanoate acting as an inhibitor of the diacylglycerol acyltransferase step. The flux-control coefficient
Aisling R Hume et al.
Journal of bacteriology, 191(24), 7554-7565 (2009-10-13)
A fatty acyl coenzyme A synthetase (FadD) from Pseudomonas putida CA-3 is capable of activating a wide range of phenylalkanoic and alkanoic acids. It exhibits the highest rates of reaction and catalytic efficiency with long-chain aromatic and aliphatic substrates. FadD
Karen M Tobin et al.
FEMS microbiology letters, 253(1), 111-118 (2005-11-02)
A number of Pseudomonas strains accumulated polyhdroxyalkanoate (PHA) from a variety of aromatic hydrocarbons. In many strains the level of PHA accumulation was dependent on the side chain length of the phenylalkanoic acid provided for growth. 4 of the 8
J G Conway et al.
Cancer research, 47(18), 4795-4800 (1987-09-15)
The diffusion of H2O2 into the cytoplasm from peroxisomes during high rates of peroxisomal beta oxidation of fatty acids was studied in perfused livers from rats treated with the hepatocarcinogenic peroxisome proliferator, nafenopin. Efflux of oxidized glutathione (GSSG) into the
Umi S Ramli et al.
The FEBS journal, 272(22), 5764-5770 (2005-11-11)
We applied metabolic control analysis to the Kennedy pathway for triacylglycerol formation in tissue cultures from the important oil crops, olive (Olea europaea L.) and oil palm (Elaeis guineensis Jacq.). When microsomal fractions were incubated at 30 degrees C rather

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service