Skip to Content
Merck
All Photos(1)

Documents

17664

Sigma-Aldrich

6-Bromo-2-naphthyl β-D-galactopyranoside

≥98.0% (HPLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H17BrO6
CAS Number:
Molecular Weight:
385.21
Beilstein:
92987
EC Number:
MDL number:
UNSPSC Code:
12352204

Assay

≥98.0% (HPLC)

optical activity

[α]20/D −45±2°, c = 1% in pyridine

impurities

≤0.1% free bromonaphthol

mp

213-216 °C

storage temp.

2-8°C

SMILES string

OC[C@H]1O[C@@H](Oc2ccc3cc(Br)ccc3c2)[C@H](O)[C@@H](O)[C@H]1O

Looking for similar products? Visit Product Comparison Guide

Other Notes

Substrate for β-galactosidase

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B Holzmann et al.
Journal of immunological methods, 60(3), 359-367 (1983-06-10)
Antibodies conjugated to enzymes such as horseradish peroxidase or alkaline phosphatase are widely used to detect antibody binding to individual cells or tissue sections through the deposit of insoluble colored reaction products. However, the presence of endogenous enzyme activity, especially
The colorimetric estimation and histochemical demonstration of beta-d-galactosidase.
R B COHEN et al.
The Journal of biological chemistry, 195(1), 239-249 (1952-03-01)
Histochemical demonstration of beta-D-galactosidase in the rat.
A M RUTENBURG et al.
The journal of histochemistry and cytochemistry : official journal of the Histochemistry Society, 6(2), 122-129 (1958-03-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service