15336
Lead ionophore II
Selectophore™
Synonym(s):
MBDiBDTC, S,S′-Methylenebis(N,N-diisobutyldithiocarbamate
About This Item
Recommended Products
grade
for ion-selective electrodes
Quality Level
product line
Selectophore™
form
solid
mp
70-71 °C
SMILES string
CC(C)CN(CC(C)C)C(=S)SCSC(=S)N(CC(C)C)CC(C)C
InChI
1S/C19H38N2S4/c1-14(2)9-20(10-15(3)4)18(22)24-13-25-19(23)21(11-16(5)6)12-17(7)8/h14-17H,9-13H2,1-8H3
InChI key
ZHJPGLVVCSNIEG-UHFFFAOYSA-N
General description
Application
Other Notes
Legal Information
related product
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service