Skip to Content
Merck
All Photos(1)

Documents

15142

Supelco

Bisphenol F bis(2,3-dihydroxypropyl) ether

analytical standard, mixture of 3 isomers ortho-ortho, ortho-para, para-para

Synonym(s):

Bis[4-(2,3-dihydroxypropoxy)phenyl]methane

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H24O6
CAS Number:
Molecular Weight:
348.39
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:

grade

analytical standard

Assay

≥95.0% (total assay of the 3 isomers, HPLC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

cleaning products
cosmetics
food and beverages
personal care

format

neat

SMILES string

OCC(O)COc1ccc(Cc2ccc(OCC(O)CO)cc2)cc1

InChI

1S/C19H24O6/c20-10-16(22)12-24-18-5-1-14(2-6-18)9-15-3-7-19(8-4-15)25-13-17(23)11-21/h1-8,16-17,20-23H,9-13H2

InChI key

VGJRHUPDMVWBFM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Other Notes

Standard for determining toxic monomers released from polymers of the inner coating of cans

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chromatographia, 34, 67-67 (1992)
P.P. Losada et al.
Fresenius Journal of Analytical Chemistry, 345, 527-527 (1993)
J.S. Gandara et al.
Journal of Chromatographic Science, 30, 11-11 (1992)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service