Skip to Content
Merck
All Photos(1)

Key Documents

11648

Sigma-Aldrich

Borax Anhydrous

anhydrous, ≥98%, granular (dry)

Synonym(s):

Sodium tetraborate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Na2B4O7
CAS Number:
Molecular Weight:
201.22
EC Number:
MDL number:
UNSPSC Code:
12352302

Quality Level

Assay

≥98%

form

granular (dry)

impurities

≤0.005% heavy metals (as Pb)

anion traces

chloride (Cl-): ≤1000 mg/kg
sulfate (SO42-): ≤500 mg/kg

cation traces

Fe: ≤50 mg/kg

SMILES string

[Na+].[Na+].[O-]B1Ob2ob([O-])ob(O1)o2

InChI

1S/B4O7.2Na/c5-1-7-3-9-2(6)10-4(8-1)11-3;;/q-2;2*+1

InChI key

UQGFMSUEHSUPRD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

replaced by

Product No.
Description
Pricing

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Repr. 1B

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service