Skip to Content
Merck
All Photos(1)

Key Documents

11619

Sigma-Aldrich

Potassium metaborate

≥31% B2O3 basis, ≥42% K2O basis

Synonym(s):

Potassium borate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KBO2 · ~1.5H2O
CAS Number:
Molecular Weight:
81.91 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.22

Quality Level

composition

B2O3, ≥31%
K2O, ≥42%

impurities

≤0.005% heavy metals (as Pb)

anion traces

chloride (Cl-): ≤500 mg/kg
sulfate (SO42-): ≤1000 mg/kg

cation traces

Fe: ≤50 mg/kg

SMILES string

[K+].[O-]B=O

InChI

1S/BO2.K/c2-1-3;/q-1;+1

InChI key

JVUYWILPYBCNNG-UHFFFAOYSA-N

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Repr. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G P Rosa et al.
Bioorganic & medicinal chemistry, 29, 115873-115873 (2020-11-27)
The human skin is constantly exposed to external factors that affect its integrity, UV radiation being one of the main stress factors. The repeated exposure to this radiation leads to increased production of Reactive Oxygen Species (ROS) which activate a
Diana I S P Resende et al.
Molecules (Basel, Switzerland), 25(12) (2020-06-21)
Antioxidants have long been used in the cosmetic industry to prevent skin photoaging, which is mediated by oxidative stress, making the search for new antioxidant compounds highly desirable in this field. Naturally occurring xanthones are polyphenolic compounds that can be

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service