Skip to Content
Merck
All Photos(1)

Key Documents

11225

Sigma-Aldrich

Ammonium sulfate

puriss., ≥99%

Synonym(s):

Mascagnite

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(NH4)2SO4
CAS Number:
Molecular Weight:
132.14
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

grade

puriss.

Assay

≥99%

impurities

≤0.001% heavy metals (as Pb)

pH

5-6 (20 °C, 5%)

mp

>280 °C (dec.) (lit.)

density

1.77 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤20 mg/kg

cation traces

As: ≤5 mg/kg
Ca: ≤50 mg/kg
Fe: ≤10 mg/kg
K: ≤50 mg/kg
Mg: ≤50 mg/kg
Na: ≤50 mg/kg
Se: ≤30 mg/kg

SMILES string

N.N.OS(O)(=O)=O

InChI

1S/2H3N.H2O4S/c;;1-5(2,3)4/h2*1H3;(H2,1,2,3,4)

InChI key

BFNBIHQBYMNNAN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

514502
CorpBase ID (for auto-filling citation data) null
D A DePasquale et al.
Applied and environmental microbiology, 56(12), 3711-3717 (1990-12-01)
In this study we examined the mechanism by which ammonium bicarbonate inhibits mycotoxigenic fungi. Elevated extracellular pH, alone, was not responsible for the antifungal activity. Although conidia of Penicillium griseofulvum and Fusarium graminearum had internal pH (pHi) values as high
Deliquescence and crystallization of ammonium sulfate particles internally mixed with water-soluble organic compounds
Parsons MT, et al.
The Journal of Physical Chemistry A, 108(52), 11600-11608 (2004)
Separation of proteins by ammonium sulfate gradient solubilization.
T P King
Biochemistry, 11(3), 367-371 (1972-02-01)
Neutron-Diffraction Study of the Structures of Ferroelectric and Paraelectric Ammonium Sulfate.
Schlemper EO, et al.
J. Chem. Phys. , 44(12), 4498-4509 (1966)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service