Skip to Content
Merck
All Photos(1)

Key Documents

08901

SAFC

Cysteamine hydrochloride

Synonym(s):

β-Mercapto­ethylamine hydrochloride, 2-Amino­ethane­thiol hydrochloride, 2-Mercapto­ethyl­amine hydrochloride, Decarboxy­cysteine hydrochloride, Thio­ethanol­amine hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HSCH2CH2NH2 · HCl
CAS Number:
Molecular Weight:
113.61
Beilstein:
3590083
EC Number:
MDL number:
UNSPSC Code:
12352200

Assay

98-102% (wt., GC)

concentration

30.6-31.8% (AT)

impurities

≤2.0% cystamine (GC)
≤20 mg/kg heavy metals
≤5000 mg/kg residual solvents (tert-butylmethyl ether
≤5000 mg/kg residual solvents (ethanol)

loss

≤1.0% loss on drying

mp

67-71 °C

solubility

H2O: 3.408 g in 30 mL, clear, colorless

cation traces

Al: ≤5 mg/kg
Ba: ≤5 mg/kg
Bi: ≤5 mg/kg
Ca: ≤10 mg/kg
Cd: ≤5 mg/kg
Co: ≤5 mg/kg
Cr: ≤5 mg/kg
Cu: ≤5 mg/kg
Fe: ≤5 mg/kg
K: ≤50 mg/kg
Li: ≤5 mg/kg
Mg: ≤5 mg/kg
Mn: ≤5 mg/kg
Mo: ≤5 mg/kg
Na: ≤50 mg/kg
Ni: ≤50 mg/kg
Pb: ≤5 mg/kg
Sr: ≤5 mg/kg
Zn: ≤5 mg/kg

storage temp.

2-8°C

SMILES string

Cl[H].NCCS

InChI

1S/C2H7NS.ClH/c3-1-2-4;/h4H,1-3H2;1H

InChI key

OGMADIBCHLQMIP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service