Skip to Content
Merck
All Photos(1)

Key Documents

08483

Supelco

Hexahydro-4-methylphthalic anhydride, mixture of cis and trans

analytical standard

Synonym(s):

4-Methyl-1,2-cyclohexanedicarboxylic anhydride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H12O3
CAS Number:
Molecular Weight:
168.19
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

Assay

≥98.0% (GC)

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

refractive index

n20/D 1.473-1.480
n20/D 1.477 (lit.)

density

1.162 g/mL at 25 °C (lit.)

application(s)

environmental

format

neat

SMILES string

CC1CCC2C(C1)C(=O)OC2=O

InChI

1S/C9H12O3/c1-5-2-3-6-7(4-5)9(11)12-8(6)10/h5-7H,2-4H2,1H3

InChI key

FKBMTBAXDISZGN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Methylhexahydrophthalic anhydride, a congener of phthalic anhydride, is utilized in the production of plastics, paints, and electronic components. It can potentially result in allergenic effects on humans upon exposure and cause immunological reactions.

Application

Hexahydro-4-methylphthalic anhydride, mixture of cis and trans may be used as an analytical standard for the determination of the analyte in human biological samples by gas chromatography-mass spectrometry (GC-MS).

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Resp. Sens. 1 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH SVHC Candidate List

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P Pfäffli et al.
The Analyst, 116(12), 1333-1336 (1991-12-01)
A gas-chromatographic technique using 63Ni electron-capture detection was applied to the determination of 4-methyl-cis-hexahydrophthalic anhydride in the blood of workers occupationally exposed to this airborne agent. The detection limit was 0.24 nmol ml-1. For occupational exposure to between 0.14 and
Marina C Jeppsson et al.
Inhalation toxicology, 21(12), 1013-1020 (2009-09-24)
Methylhexahydrophthalic anhydride (MHHPA) is a reactive, low molecular weight chemical used in products such as plastics, paints, and electronic components. Exposure to MHHPA may lead to work-related airway diseases such as rhinitis, conjunctivitis, and asthma. Twelve subjects employed at a
C H Lindh et al.
Toxicology and applied pharmacology, 153(2), 152-160 (1999-01-08)
Hexahydrophthalic anhydride (HHPA) and methylhexahydrophthalic anhydride (MHHPA) are highly allergenic compounds used in the chemical industry. The aim of this study was to characterize the protein adducts in erythrocytes following exposure to HHPA and MHHPA. Blood and urine samples were

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service