Skip to Content
Merck
All Photos(1)

Key Documents

07675

Sigma-Aldrich

4′-Aminodibenzo-18-crown-6

≥98.0% (GC)

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
855,00 kr.

855,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
855,00 kr.

About This Item

Empirical Formula (Hill Notation):
C20H25NO6
CAS Number:
Molecular Weight:
375.42
Beilstein:
8865948
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

855,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Quality Level

Assay

≥98.0% (GC)

mp

159-163 °C

functional group

ether

SMILES string

Nc1ccc2OCCOCCOc3ccccc3OCCOCCOc2c1

InChI

1S/C20H25NO6/c21-16-5-6-19-20(15-16)27-14-10-23-8-12-25-18-4-2-1-3-17(18)24-11-7-22-9-13-26-19/h1-6,15H,7-14,21H2

InChI key

FFYZYAKXQMHVQE-UHFFFAOYSA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service