Skip to Content
Merck
All Photos(1)

Key Documents

06725

Sigma-Aldrich

N-(2-Aminoethyl)-5-isoquinolinesulfonamide hydrochloride

≥97.0% (HPLC)

Synonym(s):

H-9

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H13N3O2S · HCl
CAS Number:
Molecular Weight:
287.77
Beilstein:
6548866
MDL number:
UNSPSC Code:
12352202

Assay

≥97.0% (HPLC)

solubility

methanol: 1 mg/mL, clear, colorless

SMILES string

Cl.NCCNS(=O)(=O)c1cccc2cnccc12

Biochem/physiol Actions

Selective casein kinase 1 inhibitor. Potent inhibitor of histamine release from human basophils when induced by phorbol ester or DiC8, but not when induced by anti-IgE, fMLP (formyl-Met-Leu-Phe) and calcium ionophore A23187. This implicates divergent pathways of basophil activation.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T Chijiwa et al.
The Journal of biological chemistry, 264(9), 4924-4927 (1989-03-25)
When screening various isoquinolinesulfonamide compounds which we synthesized, CKI-7, N-(2-amino-ethyl)-5-chloroisoquinoline-8-sulfonamide, was found to have a potent inhibitory action against casein kinase I and a much weaker effect on casein kinase II and other protein kinases. Kinetic analysis indicated that CKI-7
D O Thueson et al.
Biochemical and biophysical research communications, 144(2), 732-740 (1987-04-29)
The release of histamine and other inflammatory mediators from human basophils is triggered by numerous stimuli, including chemical, physical and receptor-mediated activators. Several mechanisms of cell activation including protein kinase C activation have been proposed to operate in these cells.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service