06696
3-Amino-9-ethylcarbazole
for HPLC derivatization, LiChropur™, ≥97.0% (HPLC)
Synonym(s):
9-Ethylcarbazol-3-amine, AEC
About This Item
Recommended Products
grade
derivatization grade ((HPLC))
for HPLC derivatization
Quality Level
Assay
≥97.0% (HPLC)
quality
LiChropur™
technique(s)
HPLC: suitable
storage temp.
2-8°C
SMILES string
CCn1c2ccccc2c3cc(N)ccc13
InChI
1S/C14H14N2/c1-2-16-13-6-4-3-5-11(13)12-9-10(15)7-8-14(12)16/h3-9H,2,15H2,1H3
InChI key
OXEUETBFKVCRNP-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Specificity
Legal Information
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 3 Oral - Carc. 1B
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Regulatory Listings
Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.
EU REACH Annex XVII (Restriction List)
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service