Recommended Products
description
weakly acidic
form
beads
parameter
120 °C max. temp.
moisture
47-53%
technique(s)
LPLC: suitable
loss
~45% loss on drying, 110 °C
matrix
methacrylic (gel)
matrix active group
carboxylic acid functional group
particle size
19-26 mesh
580-780 μm
operating pH
4-14
capacity
4.1 meq/mL by wetted bed volume
separation technique
cation exchange
Looking for similar products? Visit Product Comparison Guide
General description
Legal Information
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service