Skip to Content
Merck
All Photos(1)

Key Documents

06044

Sigma-Aldrich

3-(Allyloxycarbonylamino)-1-propanol

≥97.0% (GC)

Synonym(s):

Allyl N-(3-hydroxypropyl)carbamate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH2=CHCH2OCONH(CH2)3OH
CAS Number:
Molecular Weight:
159.18
Beilstein:
7579395
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥97.0% (GC)

reaction suitability

reagent type: cross-linking reagent

refractive index

n20/D 1.472

density

1.097 g/mL at 20 °C (lit.)

SMILES string

OCCCNC(=O)OCC=C

InChI

1S/C7H13NO3/c1-2-6-11-7(10)8-4-3-5-9/h2,9H,1,3-6H2,(H,8,10)

InChI key

ZKLMTPLBEYYNKM-UHFFFAOYSA-N

Other Notes

Introduction of an aminopropyl linker in a daunomycinone derivative[1]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

228.2 °F - closed cup

Flash Point(C)

109 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zoltán Dienes et al.
The Journal of organic chemistry, 61(20), 6958-6970 (1996-10-04)
The BF(3).Et(2)O-promoted Diels-Alder addition of 1-acetylvinyl RADO(Et)-ate (RADO(Et)-ate = 3-ethyl-2-oxo-6,8-dioxa-3-azabicyclo[3.2.1]octane-7-exo-carboxylate) to 1-(dimethoxymethyl)-2,3,5,6-tetramethylidene-7-oxabicyclo[2.2.1]heptane led to one major monoadduct that added to 1,2-didehydrobenzene and was converted into (-)-4-demethoxy-7-deoxydaunomycinone and (2R)-12-acetoxy-2-acetyl-5-(bromomethyl)-1,2,3,4-tetrahydronaphthacen-2-yl RADO(Et)-ate. The latter compound was used to construct (8R)-8-acetyl-6,8-dihydroxy-11-[[(3'-[(aminopropyl)oxy]-, -4'-[(aminobutyl)oxy], and -5'-[(aminopentyl)oxy]methyl]-7,8,9,10-tetrahydronaphthacene-5,12-dione

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service