Skip to Content
Merck
All Photos(1)

Key Documents

28-2100

Sigma-Aldrich

Sodium cacodylate trihydrate

≥98.0%

Synonym(s):

Cacodylic acid sodium salt trihydrate, Dimethylarsinic acid sodium salt, Dimethylarsonic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2AsO2Na · 3H2O
CAS Number:
Molecular Weight:
214.03
Beilstein:
3702348
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
Assay:
≥98.0%
form:
solid

Assay

≥98.0%

form

solid

availability

available only in Japan

pH

8.7 (50 g/L)

storage temp.

15-25°C

SMILES string

[Na+].[H]O[H].[H]O[H].[H]O[H].C[As](C)([O-])=O

InChI

1S/C2H7AsO2.Na.3H2O/c1-3(2,4)5;;;;/h1-2H3,(H,4,5);;3*1H2/q;+1;;;/p-1

InChI key

RLGWPHBPRCROJO-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Suitability

for electron microscopy

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dean Brady et al.
BMC biotechnology, 8, 8-8 (2008-02-02)
Enzymes have found extensive and growing application in the field of chemical organic synthesis and resolution of chiral intermediates. In order to stabilise the enzymes and to facilitate their recovery and recycle, they are frequently immobilised. However, immobilisation onto solid
Elisheva B Goldman et al.
Tissue engineering. Part A, 21(5-6), 1013-1023 (2014-11-05)
Impaired salivary gland (SG) function leading to oral diseases is relatively common with no adequate solution. Previously, tissue engineering of SG had been proposed to overcome this morbidity, however, not yet clinically available. Multiwall inorganic (tungsten disulfide [WS2]) nanotubes (INT-WS2)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service