Skip to Content
Merck
All Photos(1)

Key Documents

28-1840

Sigma-Aldrich

Sodium benzoate

SAJ first grade, ≥98.5%

Synonym(s):

Benzoic acid sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5COONa
CAS Number:
Molecular Weight:
144.10
Beilstein:
3572467
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
Assay:
≥98.5%
grade:
SAJ first grade
form:
solid

grade

SAJ first grade

vapor pressure

<0.01 hPa ( 20 °C)

Assay

≥98.5%

form

solid

availability

available only in Japan

pH

7.0-8.5 (25 °C, 144.1 g/L)

mp

>300 °C (lit.)

storage temp.

15-25°C

SMILES string

[Na+].[O-]C(=O)c1ccccc1

InChI

1S/C7H6O2.Na/c8-7(9)6-4-2-1-3-5-6;/h1-5H,(H,8,9);/q;+1/p-1

InChI key

WXMKPNITSTVMEF-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Saurabh Khasnavis et al.
Journal of neuroimmune pharmacology : the official journal of the Society on NeuroImmune Pharmacology, 7(2), 424-435 (2011-06-28)
DJ-1 (PARK7) is a neuroprotective protein that protects cells from oxidative stress. Accordingly, loss-of-function DJ-1 mutations have been linked with a familial form of early onset Parkinson disease. Mechanisms by which DJ-1 level could be enriched in the CNS are
Yumei Dai et al.
Journal of food science, 75(7), M482-M488 (2011-05-04)
Growth of Saccharomyces cerevisiae and Zygosaccharomyces bailii cells was monitored in the presence of sodium benzoate and eugenol alone or combined. The two antimicrobials' concentration, addition order, and timing were varied to determine and quantify any additive inhibitory effect on
Xu Huang et al.
CNS neuroscience & therapeutics, 18(1), 4-13 (2012-01-28)
D-Serine, the endogenous co-agonist of N-methyl-D-aspartate (NMDA) receptors, has been recognized as an important gliotransmitter in the mammalian brain. D-serine has been shown to prevent psychostimulant-induced decrease in hippocampal neurogenesis. However, the mechanism whereby D-serine regulates neurogenesis has not been
Jia Li et al.
Environmental science and pollution research international, 18(9), 1567-1573 (2011-05-18)
This study investigated the effect of copper on chemical oxygen demand (COD) removal efficiency and on the properties (mainly settling and dewatering) and the composition of extracellular polymeric substances (EPS) when 20 mg/L Cu(II) was continuously dosed to a sequencing
Yue Mu et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 83(1), 130-135 (2011-09-06)
The toxicity of sodium benzoate to trypsin was investigated by fluorescence spectroscopy, synchronous fluorescence spectroscopy, UV-visible absorption spectroscopy and circular dichroism (CD) spectroscopy under mimic physiological conditions. Sodium benzoate could unfold trypsin by decreasing the β-sheet structure, which leads to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service