Skip to Content
Merck
All Photos(1)

Key Documents

28-1310

Sigma-Aldrich

Silver nitrate

JIS special grade, ≥99.8%

Synonym(s):

Nitric acid silver(I) salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
AgNO3
CAS Number:
Molecular Weight:
169.87
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
Assay:
≥99.8%
grade:
JIS special grade
form:
solid
Pricing and availability is not currently available.

grade

JIS special grade

vapor density

5.8 (vs air)

Assay

≥99.8%

form

solid

availability

available only in Japan

mp

212 °C (dec.) (lit.)

storage temp.

15-25°C

SMILES string

[O-][N+]([O-])=O.[Ag+]

InChI

1S/Ag.NO3/c;2-1(3)4/q+1;-1

InChI key

SQGYOTSLMSWVJD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Silver nitrate is a versatile reagent that can be prepared by reacting silver with nitric acid. It is used in various fields such as organic synthesis, photography, silver paints, dyes, chemical analysis, gravimetric analysis, and pharmaceutical preparations.

Application

Silver nitrate can be used as a catalyst in the:
  • [3+2]-Cycloaddition of Ͱ-diazocarbonyl compounds with arenediazonium salts to synthesize 2,5-disubstituted tetrazoles.
  • Oxidation of aromatic, aliphatic, and conjugated aldehydes to corresponding carboxylic acids in the presence of H2O2 as an oxidant.
  • Lactamization of methyl Ͱ,Ͱ-disubstituted Ͱ-isocyanoacetates with primary amines to synthesize 3,5,5-trisubstituted imidazolones.

It can also be used as a:
  • Reagent to synthesize bisazetanes from the oxidation of azetidine.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Met. Corr. 1 - Ox. Sol. 2 - Repr. 1B - Skin Corr. 1A

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Liming Wang et al.
ACS nano, 9(6), 6532-6547 (2015-05-23)
To predict potential medical value or toxicity of nanoparticles (NPs), it is necessary to understand the chemical transformation during intracellular processes of NPs. However, it is a grand challenge to capture a high-resolution image of metallic NPs in a single
Patrycja Bober et al.
Biomacromolecules, 15(10), 3655-3663 (2014-08-28)
In this work, flexible and free-standing composite films of nanofibrillated cellulose/polypyrrole (NFC/PPy) and NFC/PPy-silver nanoparticles (NFC/PPy-Ag) have been synthesized for the first time via in situ one-step chemical polymerization and applied in potential biomedical applications. Incorporation of NFC into PPy
Gownolla Malegowd Raghavendra et al.
Carbohydrate polymers, 93(2), 553-560 (2013-03-19)
Natural carbohydrates (polysaccharides): gum acacia (GA) and gaur gum (GG) were employed in dilute solutions: 0.3%, 0.5% and 0.7% (w/v), as effective reductants for the green synthesis of silver nanoparticles (AgNPs) from AgNO3. The formed AgNPs were impregnated into cellulose
Lara Settimio et al.
Environmental pollution (Barking, Essex : 1987), 191, 151-157 (2014-05-20)
The fate and lability of added soluble Ag in soils over time was examined by measurement of labile metal (E-value) by isotopic dilution using the (110m)Ag radioactive isotope and the solid-phase speciation of Ag by X-ray absorption near edge structure
Ali Hebeish et al.
Carbohydrate polymers, 92(1), 407-413 (2012-12-12)
Innovative CMC-based hydrogels with great potentials for usage in medical area were principally synthesized as per two strategies .The first involved reaction of epichlorohydrin in alkaline medium containing silver nitrate to yield silver nano-particles (AgNPs)-loaded CMC hydrogel. While CMC acted

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service