Skip to Content
Merck
All Photos(1)

Documents

24-2740

Sigma-Aldrich

Phosphorus pentachloride

CP

Synonym(s):

Phosphorus(V) chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
PCl5
CAS Number:
Molecular Weight:
208.24
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:

grade

CP

form

solid

availability

available only in Japan

pH

1 (5 g/L)

SMILES string

ClP(Cl)(Cl)(Cl)Cl

InChI

1S/Cl5P/c1-6(2,3,4)5

InChI key

UHZYTMXLRWXGPK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B - STOT RE 2 Inhalation

Target Organs

Respiratory Tract

Supplementary Hazards

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xinlin Du et al.
Analytical biochemistry, 372(2), 213-221 (2007-10-30)
A method to determine 18 O kinetic isotope effects (KIEs) in the hydrolysis of GTP that is generally applicable to reactions involving other nucleotide triphosphates is described. Internal competition, where the substrate of the reaction is a mixture of 18

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service