Skip to Content
Merck
All Photos(1)

Documents

18-0875

Sigma-Aldrich

Lead stain solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12000000

form

liquid

availability

available only in Japan

storage temp.

2-8°C

Suitability

for electron microscopy

Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Lact. - Met. Corr. 1 - Repr. 1A - Skin Irrit. 2

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tomohiro Matsumoto et al.
Diabetologia, 62(10), 1915-1927 (2019-07-19)
To treat obesity and related diseases, considerable effort has gone into developing strategies to convert white adipocytes into thermogenic brown-like adipocytes ('browning'). The purpose of this study was to identify the most efficient signal control for browning. To identify the
Yuka Suzuki et al.
Scientific reports, 9(1), 8461-8461 (2019-06-13)
Reformation of a functional nucleus at the end of mitosis is crucial for normal cellular activity. Reconstitution approaches using artificial beads in frog egg extracts have clarified the molecules required for nuclear formation in vitro. However, the spatiotemporal regulation of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service