Skip to Content
Merck
All Photos(1)

Key Documents

13-1980

Sigma-Aldrich

4-Methoxyphenol

SAJ first grade, ≥97.0%

Synonym(s):

4-Hydroxyanisole, 4-MP, HQMME, Hydroquinone monomethyl ether, MEHQ, p-Guaiacol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H4OH
CAS Number:
Molecular Weight:
124.14
Beilstein:
507924
EC Number:
MDL number:
UNSPSC Code:
12352104
PubChem Substance ID:

grade

SAJ first grade

vapor density

4.3 (vs air)

vapor pressure

<0.01 mmHg ( 20 °C)

Assay

≥97.0%

autoignition temp.

789 °F

availability

available only in Japan

bp

243 °C (lit.)

mp

55-57 °C (lit.)

SMILES string

COc1ccc(O)cc1

InChI

1S/C7H8O2/c1-9-7-4-2-6(8)3-5-7/h2-5,8H,1H3

InChI key

NWVVVBRKAWDGAB-UHFFFAOYSA-N

Gene Information

human ... TYR(7299)

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Byeoung-Soo Park et al.
Journal of agricultural and food chemistry, 51(1), 295-300 (2002-12-28)
Volatiles were isolated from the dried inner bark of Tabebuia impetiginosa using steam distillation under reduced pressure followed by continuous liquid-liquid extraction. The extract was analyzed by gas chromatography and gas chromatography-mass spectrometry. The major volatile constituents of T. impetiginosa
Jean Paul Ortonne et al.
Cutis, 74(4), 261-264 (2004-11-24)
The objective of this open-label, noncontrolled study was to evaluate the safety of a combination solution containing 4-hydroxyanisole (mequinol) 2%/tretinoin 0.01% (Solagé) with a sunscreen in the treatment of solar lentigines. The study included a total of 406 subjects for
Lisa M Colosi et al.
Environmental science & technology, 41(3), 891-896 (2007-03-03)
Natural organic matter (NOM) of hydroxylated aromatic character can undergo catalyst-mediated self-coupling reactions to form larger molecular aggregates. Indeed, such reactions are central to natural humification processes. Nonhydroxylated persistent aromatic contaminants such as polychlorinated biphenyls (PCBs) are, conversely, inert with
Majid Y Moridani
Cancer letters, 243(2), 235-245 (2006-01-24)
In the current work we investigated for the first time the biochemical basis of 4-hydroxyanisole (4-HA) induced toxicity in B16-F0 melanoma cells. It was found that dicoumarol, a diaphorase inhibitor, and 1-bromoheptane, a GSH depleting agent, increased 4-HA induced toxicity
Yoshinori Kadoma et al.
Molecules (Basel, Switzerland), 15(3), 1103-1112 (2010-03-26)
Compoundswith two phenolic OH groups like curcumin possess efficient antioxidant and anti-inflammatory activity. We synthesized p-cresol dimer (2,2'-dihydroxy-5,5'-dimethylbiphenol, 2a) and p-methoxyphenol dimer (2,2'-dihydroxy-5,5'-dimethoxybiphenol, 2b) by ortho-ortho coupling reactions of the parent monomers, p-cresol (1a) and p-methoxyphenol (1b), respectively. Their antioxidant

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service