Skip to Content
Merck
All Photos(1)

Key Documents

03-0270

Sigma-Aldrich

Barium chloride

SAJ first grade, ≥98.0%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BaCl2
CAS Number:
Molecular Weight:
208.23
EC Number:
MDL number:
UNSPSC Code:
12352300

grade

SAJ first grade

Assay

≥98.0%

availability

available only in Japan

mp

963 °C (lit.)

density

3.856 g/mL at 25 °C (lit.)

SMILES string

Cl[Ba]Cl

InChI

1S/Ba.2ClH/h;2*1H/q+2;;/p-2

InChI key

WDIHJSXYQDMJHN-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Inhalation - Eye Irrit. 2

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sarah E Pierce et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 15(42), 11244-11255 (2009-09-12)
Isoguanine (2-oxo-6-amino-guanine), a natural but non-standard base, exhibits unique self-association properties compared to its isomer, guanine, and results in formation of different higher order DNA structures. In this work, the higher order structures formed by oligonucleotides containing guanine repeats or
Anne R Crecelius et al.
American journal of physiology. Heart and circulatory physiology, 307(5), H782-H791 (2014-06-29)
We tested the hypothesis that activation of inwardly rectifying potassium (KIR) channels and Na(+)-K(+)-ATPase, two pathways that lead to hyperpolarization of vascular cells, contributes to both the onset and steady-state hyperemic response to exercise. We also determined whether after inhibiting
Michael Xavier Doss et al.
PloS one, 7(7), e40288-e40288 (2012-07-21)
Human induced pluripotent stem cell-derived cardiomyocytes (hiPSC-CM) hold promise for therapeutic applications. To serve these functions, the hiPSC-CM must recapitulate the electrophysiologic properties of native adult cardiomyocytes. This study examines the electrophysiologic characteristics of hiPSC-CM between 11 and 121 days
Chiara Callies et al.
Journal of cell science, 124(Pt 11), 1936-1942 (2011-05-12)
The stiffness of vascular endothelial cells is crucial to mechanically withstand blood flow and, at the same time, to control deformation-dependent nitric oxide release. However, the regulation of mechanical stiffness is not yet understood. There is evidence that a possible
Mirna González-González et al.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences, 893-894, 182-186 (2012-03-27)
Recovery and purification of stem cells are determining steps in order to obtain the purity and viability required for transplantation. In this context, immunochemical techniques have been widely preferred due to their high selectivity. CD133, a glycoprotein expressed by stem

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service