Skip to Content
Merck
All Photos(1)

Key Documents

01-1410

Sigma-Aldrich

Activated carbon

SAJ first grade

Synonym(s):

Activated charcoal, Charcoal activated

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C
CAS Number:
Molecular Weight:
12.01
EC Number:
MDL number:
UNSPSC Code:
23201100
PubChem Substance ID:
Pricing and availability is not currently available.

grade

SAJ first grade

vapor pressure

<0.1 mmHg ( 20 °C)

form

powder, crystals or granules

autoignition temp.

842 °F

availability

available only in Japan

resistivity

1375 μΩ-cm, 20°C (graphite)

mp

3550 °C (lit.)

SMILES string

[C]

InChI

1S/C

InChI key

OKTJSMMVPCPJKN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Timothy J Fahey et al.
Ecological applications : a publication of the Ecological Society of America, 23(5), 1185-1201 (2013-08-24)
To examine the mechanisms of earthworm effects on forest soil C and N, we double-labeled leaf litter with 13C and 15N, applied it to sugar maple forest plots with and without earthworms, and traced isotopes into soil pools. The experimental
Nathalie Butt et al.
Ecological applications : a publication of the Ecological Society of America, 23(4), 936-943 (2013-07-20)
A typical way to quantify aboveground carbon in forests is to measure tree diameters and use species-specific allometric equations to estimate biomass and carbon stocks. Using "citizen scientists" to collect data that are usually time-consuming and labor-intensive can play a
Energy policy: A low-carbon road map for China.
Zhu Liu et al.
Nature, 500(7461), 143-145 (2013-08-09)
Elisangela Heiderscheidt et al.
Journal of environmental sciences (China), 25(4), 659-669 (2013-08-09)
The drainage of peatland areas for peat extraction, agriculture or bioenergy requires affordable, simple and reliable treatment methods that can purify waters rich in particulates and dissolved organic carbon. This work focused on the optimisation of chemical purification process for
Abstracts of the 9th International Conference On Homocysteine and One-Carbon Metabolism - HCY2013. Dublin, Ireland. September 8-12, 2013.
Journal of inherited metabolic disease, 36 Suppl 1, S1-54 (2013-07-25)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service