ZR0L52220
Milli-Q® HR High-Flow RO Water Purification System
Central water purification system for up to 4400 L/day Type 3 water.
Synonym(s):
Milli-Q® HR 7220 High-Flow RO Water Purification System
About This Item
Recommended Products
AC/DC input
100 V / 230 V, 50-60 Hz
Quality Level
product line
Milli-Q® HR
feature
Ionic rejection by RO: 95 - 99%
Organic rejection by RO: >=99% for MW >= 200
packaging
pkg of 1 unit
parameter
103 kg operating weight (252 lb)
220 L/hr make-up flow rate
4400 L/day max. usage
system depth × H × W
54.2 cm (21.3 in.) × 124 cm (48.8 in.) × 54.3 cm (21.4 in.)
input
feed water nature potable tap water
output
product water quality: type 3 water (RO water)
Related Categories
General description
Milli-Q® HR 7000 series systems can be integrated into a centralized system, providing total control of all parameters within the system itself as well as within the external pure water distribution loop.
These central systems are embedded with our unique MyMilli-Q™ Remote Care online monitoring and service capability, which operates as an extension of Milli-Q® Services. This digital service lets you securely and remotely manage your system, supporting the lab′s optimal productivity.
Application
It can feed instruments, including dishwashers, autoclaves, humidity and environmental chambers, and ice machines.
It can also feed Milli-Q® ultrapure water purification systems.
Features and Benefits
- Enhances sustainability and reduces running costs, making your facility more productive, environmentally friendly, and cost-effective.
- Superior ergonomic design for quick and easy maintenance.
- Progard® Pretreatment cartridges remove particles, free chlorine and colloids.
- An interactive touchscreen with helpful wizards for ease of operation and maintenance.
- Modern data management capabilities for greater accessibility and data traceability.
- MyMilli-Q™ Remote Care monitoring and service capability.
- Equipped with patented E.R.A.® (Evolutive Reject Adjustment) technology to save you water, time and money. It automatically assesses feed water quality to produce a constant flow rate and optimize water recovery (between 45 and 75%).
- Compatible with SDS 500 storage, protection, and distribution system, for high storage and distribution flow possibilities.
Components
- The indicated price currently corresponds to the above catalog number only; it is subject to change at any time without notice.
- This catalog number does not include consumables, accessories, and other products and/or services that may be required for the installation and the correct operation of the water purification unit over time.
- Because our customers each have specific purified water needs, we recommend that you have your purchase validated by one of our specialists. We remain at your disposal to advise you, do not hesitate to contact us!
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Experience efficient Milli-Q® HR 7000 Central RO Water Purification Systems. A more sustainable high-flow pure water system with versatile configurations and modern control. Advanced RO technology saves water. Online monitoring gives peace of mind. Request information now!
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service