ZLXM0D070
Elix® Electrodeionization (EDI) Module
Water Purification by Electrodeionization (EDI) at 70 L/hr
Select a Size
43 56 59 19.
This item is not able to be purchased as a guest user. For further information, please contact us atSelect a Size
About This Item
43 56 59 19.
This item is not able to be purchased as a guest user. For further information, please contact us atRecommended Products
material
resin (Mixed bed ion-exchange resin)
feature
removes remaining ions by electrodeionization
packaging
pkg of 1 unit
manufacturer/tradename
Elix®
parameter
70 L/hr flow rate
compatibility
for use with Elix®
Looking for similar products? Visit Product Comparison Guide
General description
Application
Features and Benefits
- Genuine replacement part for the Elix® EDI Module - ZLXM0D070.
- Contains mixed bed ion-exchange resin.
- Efficiently removes remaining ions from RO-purified water to produce constant-quality pure water, regardless of feed water quality (conductivity, CO2 levels) or RO cartridge performance.
- Eliminates the need for hazardous chemical regeneration procedures, replacement of costly resins, changing DI cartridges, or adding softeners.
- Reduces maintenance time, ensuring low and predictable running costs.
Legal Information
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service