Skip to Content
Merck
All Photos(1)

Key Documents

ZLXL000PR

Milli-Q

Feed Water Pressure Regulator

Regulates the feed water pressure from 0-25 bars, Ensures accurate water level sensing to prevent overflow for use with high flow water systems

Synonym(s):

XL Feed Pressure Regulator 0-25 bars

Sign Into View Organizational & Contract Pricing

Select a Size

1 EA
2.840,00 kr.

2.840,00 kr.


Available to ship on11 April 2025Details


Request more information

Select a Size

Change View
1 EA
2.840,00 kr.

About This Item

UNSPSC Code:
41104200
NACRES:
JA.13

2.840,00 kr.


Available to ship on11 April 2025Details


Request more information

packaging

pkg of 1 unit

compatibility

for use with Milli-Q® CLX

Looking for similar products? Visit Product Comparison Guide

General description

This feed water pressure regulator adjusts the inlet water pressure upstream of water purification systems to regulate the pressure according to the system′s requirements.

Application

For high volume water systems.

Legal Information

Milli-Q is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service