Skip to Content
Merck
All Photos(1)

Documents

SYMBSCA01

Millipore

Steritest® Symbio Shipping Case

Portable Shipping Case for Steritest® Symbio Pumps

Synonym(s):

Isolator Pump Case, Portable Case, Shipping Case

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151818
NACRES:
JA.12

material

polyethylene (Foam)
polypropylene copolymer case

manufacturer/tradename

Steritest®

L × W × H

630 mm × 492 mm × 352 mm

application(s)

pharmaceutical

compatibility

for use with Steritest® Symbio FLEX Pump Kit, 2 media (SYMBFLE01)
for use with Steritest® Symbio ISL Pump Kit, 2 media (SYMBISL01)
for use with Steritest® Symbio LFH Pump Kit (SYMBLFH01)
for use with Steritest® Symbio Pumps

General description

Steritest® Symbio Shipping Case is designed for safe and convenient transportation of the Steritest® Symbio pump, e.g. for the yearly maintenance. It is compatible with all Steritest® Symbio pump.

Legal Information

STERITEST is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service