Skip to Content
Merck
All Photos(2)

Key Documents

SYMBDTC03

Millipore

Steritest® Symbio 3 Media Drain Tray Container

Streamlines the workflow, Increases Safety

Synonym(s):

Outlet

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
11.770,00 kr.

11.770,00 kr.


Please contact Customer Service for Availability

Request more information

Select a Size

Change View
1 EACH
11.770,00 kr.

About This Item

UNSPSC Code:
23151818
NACRES:
NB.13

11.770,00 kr.


Please contact Customer Service for Availability

Request more information

material

polyphenylsulfone (PPSU) container (Drain Tray Support)

Agency

complies with EP (2.6.1)
complies with JP (4.06)
complies with USP (71)

manufacturer/tradename

Steritest®

application(s)

pharmaceutical
sterility testing

compatibility

for use with Steritest® Symbio Pump

shipped in

ambient

General description

Steritest® Symbio plastic fluid collector for 3 canisters drain tray.

Application

Accessories for Streamlining Your Workflow and Increasing Safety during sterility testing.

Legal Information

STERITEST is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service