Skip to Content
Merck
All Photos(1)

Key Documents

SX0060

Supelco

Salicylic Acid

Meets ACS Specifications GR ACS

Synonym(s):

Salicylic Acid, 2-Hydroxybenzoic Acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4(OH)COOH
CAS Number:
MDL number:
UNSPSC Code:
12352106
EC Index Number:
200-712-3

vapor pressure

<1 hPa ( 100 °C)

Quality Level

Assay

≥99%

form

crystals

technique(s)

tissue culture: suitable (plant)

ign. residue

≤0.01%

color

white

pH

2.4 ( in H2O, saturated solution)

bp

211 °C/1013 hPa

mp

157-159 °C

transition temp

flash point 157 °C

solubility

2 g/L

density

1.44 g/cm3 at 20 °C

anion traces

chloride (Cl-): ≤0.001%
sulfate (SO42-): ≤0.003%

cation traces

Fe: ≤2 ppm
heavy metals (as Pb): ≤5 ppm

shipped in

ambient

storage temp.

room temp

InChI

1S/C7H6O3/c8-6-4-2-1-3-5(6)7(9)10/h1-4,8H,(H,9,10)

InChI key

YGSDEFSMJLZEOE-UHFFFAOYSA-N

Application

Meets ACS Specifications GR ACS

Analysis Note

Assay99.0% minChloride (Cl)0.001% maxColorWhiteFormCrystalsHeavy metals (as Pb)5ppm maxIron (Fe)2ppm maxMelting point158.0-161.0°CResidue after ignition0.01% maxSubstances darkened by sulfuric acidTo pass testSulfate (SO4)0.003% max

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Repr. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

314.6 °F - closed cup

Flash Point(C)

157 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service