Skip to Content
Merck
All Photos(1)

Documents

SPGPM10RJ

Millipore

Millipore® Steripak Pressure Filter

pore size 0.22 μm, polyethersulfone membrane, 2-stack, pack of 3 ea

Synonym(s):

Steripak-GP 0.22 μm, polyethersulfone, 2-Stack sterilized by Gamma irradiation, Steripak-GP Pressure Filter Unit, pressure filtration unit, pressure sterile filter unit, pressure sterile filtering unit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104914
eCl@ss:
32031690
NACRES:
NB.22

material

polyethersulfone membrane
styrene-acrylonitrile (SAN)

Quality Level

sterility

sterile

product line

Steripak

feature

holdup volume 2.7 mL
hydrophilic

packaging

pack of 3 ea

parameter

0.35 bar max. inlet pressure (5 psi) (Reverse)
45 °C max. temp.
5.2 bar max. inlet pressure (75 psi) (Forward)

technique(s)

sterile filtration: suitable

H

76 mm

diam.

9.9 cm

filter diam.

80 mm

filtration area

100 cm2

matrix

Express PLUS

pore size

0.22 μm pore size

fitting

6 mm (1/4 in.) inlet stepped hose barb
outlet filling bell
6 mm (1/4 in.) outlet stepped hose barb

shipped in

ambient

General description

Steripak-GP filter units are designed for larger scale pressure-driven filtration of tissue culture media with or without serum. They are single use and are not autoclavable. The filter units come in two sizes, with the smaller unit able to filter up to 10 L and the larger up to 20 L. Both sizes utilize the low protein binding, fast flow 0.22 µm Millipore Express membrane. Each unit has an integral filling bell for bottle-top use and a manually operated upstream air vent to prevent membrane air locking. They are supplied sterile and ready to connect to a pump or pressure vessel (not included).

Steripak-GP filter units are designed for larger scale, pressure-driven filtration of tissue culture media with or without serum. Each filter is for a single use and is not autoclavable. The filter units come in two sizes with the smaller unit able to filter up to 10 L and the larger up to 20 L.

Features & Benefits:
- Millipore Express PLUS membrane for fastest flow rates and low protein binding
- Larger scale, pressure-driven filtration device for faster flow rates speeds time-consuming, large-volume filtrations
- 10 L and 20 L capacity sizes for maximum laboratory-scale filtration options
- Manually operated top air vent to prevent air lock stop of flow
- Filling bell for cleaner filling environment around the opening of the receptacle bottle

Applications:
Sterilization of Aqueous Solutions, Media Preparation, Buffer Preparation

Application

For Research Use Only

Principle

Pressure

Physical form

Color Code: Clear inlet/blue outlet

Preparation Note

Sterilization Method
Gamma irradiation

Other Notes

Directions for Use
  • Organism Retention: Microorganisms
  • Mode of Action: Filtering
  • Application: Sterile Filtration
  • Intended Use: Sterilization of Aqueous Solutions, Media Preparation, Buffer Preparation
  • Instructions for Use: Please refer to Instructions for Use section of Steripak user guide
  • Storage Statement: Store in dry location
  • Disposal Statement: Dispose of in accordance with applicable federal, state and local regulations.

Legal Information

EXPRESS PLUS is a trademark of Merck KGaA, Darmstadt, Germany
Millipore is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service