Skip to Content
Merck
All Photos(1)

Documents

SLM-245

Sigma-Aldrich

Minimum Essential Medium Eagle complete medium

With non-essential amino acids, ʟ-glutamine and 10% FBS, without sodium pyruvate, sterile liquid, suitable for cell culture

Synonym(s):

EMEM, MEM

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
eCl@ss:
32160801
NACRES:
NA.75

product name

MEM Complete Medium, with NEAA, 2mM L-Glut and 10% FBS, contains non-essential amino acids, 2mM L-glutamine and 10% U.S. origin FBS

Quality Level

form

liquid

technique(s)

cell culture | mammalian: suitable

Quality

pH= 7.2-7.5
Osmolality= 281-311 mOsm/kg
Endotoxin= ≤2 EU/mL
Mycoplasma= Negative
Sterility Tested= Negative for Growth
Appearance= Orange/ Red Clear Liquid
Cell Line Tested= HepG2 liver hepatocellular carcinoma

Other Notes

Concentration: Please refer to lot specific datasheet.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service